accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 40241-78-9

IUPAC Name: hydrogen 2-(methylsulfanyl)-4,5-dihydro-1H-imidazole bromide
Synonyms: hydrogen 2-(methylsulfanyl)-4,5-dihydro-1H-imidazole bromide
Molecular Weight (g/mol): 197.09
Molecular Formula: C4H9BrN2S
InChi Key: UPISCLUJZRQPPJ-UHFFFAOYSA-N
SMILES: [H+].[Br-].CSC1=NCCN1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results