accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NH2NOOFFF

CAS RN 402-14-2

IUPAC Name: 2-nitro-5-(trifluoromethyl)aniline
Synonyms: 2-nitro-5-(trifluoromethyl)aniline
Molecular Weight (g/mol): 206.12
Molecular Formula: C7H5F3N2O2
InChi Key: AUTLVHYEAAAKNM-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1[N+]([O-])=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)