accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 402-07-3

IUPAC Name: 1-bromo-2-methoxy-4-(trifluoromethyl)benzene
Synonyms: 1-bromo-2-methoxy-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 255.03
Molecular Formula: C8H6BrF3O
InChi Key: FQLLPTWRKUGXEN-UHFFFAOYSA-N
SMILES: COC1=C(Br)C=CC(=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results