accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OOS

CAS RN 40133-07-1

IUPAC Name: 4,5,6,7-tetrahydro-1-benzothiophene-2-carboxylate
Synonyms: 4,5,6,7-tetrahydro-1-benzothiophene-2-carboxylate
Molecular Weight (g/mol): 181.23
Molecular Formula: C9H9O2S
InChi Key: ROLXOQXKNDKXTA-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC2=C(CCCC2)S1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)