CAS RN 400803-05-6
CAS RN 400803-05-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Boc-4-(2-chloro-6-nitrophenyl)piperazine, 97%, Thermo Scientific™
CAS: 400803-05-6 Synonym: tert-Butyl 4-(2-chloro-6-nitrophenyl)-piperazine-1-carboxylate; 4-(2-Chloro-6-nitrophenyl)-piperazine-1-carboxylic acid tert-butyl ester