accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 400607-04-7

Br(?)

CAS RN 400607-04-7

IUPAC Name: 9-bromo-10-(naphthalen-1-yl)anthracene
Synonyms: 9-bromo-10-(naphthalen-1-yl)anthracene
Molecular Weight (g/mol): 383.29
Molecular Formula: C24H15Br
InChi Key: SYACRXBYRNYMLN-UHFFFAOYSA-N
SMILES: BrC1=C2C=CC=CC2=C(C2=CC=CC=C12)C1=CC=CC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results