accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 400-73-7

H3COClFFF

CAS RN 400-73-7

IUPAC Name: 1-chloro-4-methoxy-2-(trifluoromethyl)benzene
Synonyms: 1-chloro-4-methoxy-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 210.58
Molecular Formula: C8H6ClF3O
InChi Key: HUFKNQCAJLSTLH-UHFFFAOYSA-N
SMILES: COC1=CC=C(Cl)C(=C1)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results