accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3998-88-7

H3COONClCH3

CAS RN 3998-88-7

IUPAC Name: ethyl 2-chloro-6-methylpyridine-4-carboxylate
Synonyms: ethyl 2-chloro-6-methylpyridine-4-carboxylate
Molecular Weight (g/mol): 199.63
Molecular Formula: C9H10ClNO2
InChi Key: UMAAIDIASQFLTF-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(C)=NC(Cl)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results