CAS RN 39969-26-1
CAS RN 39969-26-1
IUPAC Name:
1-[2-(4-methoxyphenyl)ethynyl]-4-propylbenzene
Synonyms:
1-[2-(4-methoxyphenyl)ethynyl]-4-propylbenzene
Molecular Weight (g/mol):
250.34
Molecular Formula:
C18H18O
InChi Key:
NPCKVHHCQBJJIS-UHFFFAOYSA-N
SMILES:
CCCC1=CC=C(C=C1)C#CC1=CC=C(OC)C=C1
1
–
1
of
1
results
1
1-[(4-Methoxyphenyl)ethynyl]-4-n-propylbenzene, 99+%, Thermo Scientific™
CAS: 39969-26-1 Molecular Formula: C18H18O Molecular Weight (g/mol): 250.341 MDL Number: MFCD04038795 InChI Key: NPCKVHHCQBJJIS-UHFFFAOYSA-N Synonym: 1-methoxy-4-4-propylphenyl ethynyl benzene,1-2-4-methoxyphenyl ethynyl-4-propylbenzene,1-4-methoxyphenyl ethynyl-4-propylbenzene,1-4-methoxyphenyl ethynyl-4-n-propylbenzene,benzene, 1-4-methoxyphenyl ethynyl-4-propyl,1-4-methoxyphenyl-2-4-n-propylphenyl acetylene,1-2-4-methoxyphenyl-1-ethynyl-4-propylbenzene,acmc-1ae6b PubChem CID: 22908548 IUPAC Name: 1-methoxy-4-[2-(4-propylphenyl)ethynyl]benzene SMILES: CCCC1=CC=C(C=C1)C#CC2=CC=C(C=C2)OC