accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 39911-29-0

NONOO

CAS RN 39911-29-0

IUPAC Name: 3-nitro-4-(piperidin-1-yl)benzaldehyde
Synonyms: 3-nitro-4-(piperidin-1-yl)benzaldehyde
Molecular Weight (g/mol): 234.26
Molecular Formula: C12H14N2O3
InChi Key: BNNMEAFYZHALEC-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(C=CC(C=O)=C1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results