CAS RN 39905-57-2
IUPAC Name:
4-(hexyloxy)aniline
Synonyms:
4-hexyloxyaniline
Molecular Weight (g/mol):
193.29
Molecular Formula:
C12H19NO
InChi Key:
DJRKHTCUXRGYEU-UHFFFAOYSA-N
SMILES:
CCCCCCOC1=CC=C(N)C=C1
1
–
1
of
1
results
1
4-n-Hexyloxyaniline, 99%
CAS: 39905-57-2 Molecular Formula: C12H19NO Molecular Weight (g/mol): 193.29 MDL Number: MFCD00007868 InChI Key: DJRKHTCUXRGYEU-UHFFFAOYSA-N Synonym: 4-hexyloxyaniline,4-hexyloxy aniline,p-hexyloxyaniline,benzenamine, 4-hexyloxy,4-n-hexyloxyaniline,p-hexyloxy aniline,4-hexyloxy benzenamine,unii-vmx449yp3l,aniline, p-hexyloxy,vmx449yp3l PubChem CID: 38360 IUPAC Name: 4-hexoxyaniline SMILES: CCCCCCOC1=CC=C(N)C=C1