accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OONH2NOO

CAS RN 3987-92-6

IUPAC Name: methyl 4-amino-3-nitrobenzoate
Synonyms: methyl 4-amino-3-nitrobenzoate
Molecular Weight (g/mol): 196.16
Molecular Formula: C8H8N2O4
InChi Key: HNTLUEZVPLRQEV-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=C(N)C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results