accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COClOOH3C

CAS RN 39828-35-8

IUPAC Name: 2,4-dimethoxybenzoyl chloride
Synonyms: 2,4-dimethoxybenzoyl chloride
Molecular Weight (g/mol): 200.62
Molecular Formula: C9H9ClO3
InChi Key: KVIUXRJCBBXEGJ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C(Cl)=O)C(OC)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results