accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 397843-80-0

BNHOHOOH

CAS RN 397843-80-0

IUPAC Name: (4-benzamidophenyl)boronic acid
Synonyms: 4-benzamidophenylboronic acid
Molecular Weight (g/mol): 241.05
Molecular Formula: C13H12BNO3
InChi Key: FWZVIUZIYYIKRK-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(NC(=O)C2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results