accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BOHNH3CCH3HOOH

CAS RN 397843-67-3

IUPAC Name: {4-[(propan-2-yl)carbamoyl]phenyl}boronic acid
Synonyms: 4-(isopropylcarbamoyl)phenylboronic acid
Molecular Weight (g/mol): 207.04
Molecular Formula: C10H14BNO3
InChi Key: GBCSEYKTZAKRMT-UHFFFAOYSA-N
SMILES: CC(C)NC(=O)C1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results