accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CCH3H3COOONH2NH2HCl

CAS RN 39741-62-3

IUPAC Name: tert-butyl 2-amino-4-carbamoylbutanoate hydrochloride
Synonyms: tert-butyl 2-amino-4-carbamoylbutanoate hydrochloride
Molecular Weight (g/mol): 238.71
Molecular Formula: C9H19ClN2O3
InChi Key: ZEPNUNKDKQACNC-UHFFFAOYNA-N
SMILES: Cl.CC(C)(C)OC(=O)C(N)CCC(N)=O
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results