accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NH2ON

CAS RN 39640-62-5

IUPAC Name: 2-(pyridin-4-yl)acetamide
Synonyms: 4-pyridineacetamide
Molecular Weight (g/mol): 136.15
Molecular Formula: C7H8N2O
InChi Key: YEALAJQPOVKTOH-UHFFFAOYSA-N
SMILES: NC(=O)CC1=CC=NC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results