accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3959-23-7

OHOSOO

CAS RN 3959-23-7

IUPAC Name: 2-(benzenesulfonyl)acetic acid
Synonyms: (benzenesulfonyl)acetic acid
Molecular Weight (g/mol): 200.21
Molecular Formula: C8H8O4S
InChi Key: YTEFAALYDTWTLB-UHFFFAOYSA-N
SMILES: OC(=O)CS(=O)(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results