accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3ONHNOOFFF

CAS RN 395-68-6

IUPAC Name: N-[4-nitro-2-(trifluoromethyl)phenyl]acetamide
Synonyms: N-[4-nitro-2-(trifluoromethyl)phenyl]acetamide
Molecular Weight (g/mol): 248.16
Molecular Formula: C9H7F3N2O3
InChi Key: RBEVUHKSMGYLIF-UHFFFAOYSA-N
SMILES: CC(=O)NC1=C(C=C(C=C1)[N+]([O-])=O)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results