accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3945-18-4

CH3OO(S)O(R)OH3CO

CAS RN 3945-18-4

IUPAC Name: (3S,4R)-3-(acetyloxy)-3,4-dihydro-2H-pyran-4-yl acetate
Synonyms: (3S,4R)-3-(acetyloxy)-3,4-dihydro-2H-pyran-4-yl acetate
Molecular Weight (g/mol): 200.19
Molecular Formula: C9H12O5
InChi Key: OWKCFBYWQGPLSJ-BDAKNGLRSA-N
SMILES: CC(=O)O[C@H]1COC=C[C@H]1OC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results