accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 39267-79-3

OBr

CAS RN 39267-79-3

IUPAC Name: 3-bromooxetane
Synonyms: 3-bromooxetane
Molecular Weight (g/mol): 136.98
Molecular Formula: C3H5BrO
InChi Key: SZTIZZFKWQWSSP-UHFFFAOYSA-N
SMILES: BrC1COC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results