accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OOOH(E/Z)

CAS RN 39244-10-5

IUPAC Name: 3-(furan-3-yl)prop-2-enoic acid
Synonyms: 3-(furan-3-yl)prop-2-enoic acid
Molecular Weight (g/mol): 138.12
Molecular Formula: C7H6O3
InChi Key: JHAPZUDWRRBZHZ-UHFFFAOYSA-N
SMILES: OC(=O)C=CC1=COC=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results