accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3921-01-5

NNBrBr

CAS RN 3921-01-5

IUPAC Name: 2,4-dibromopyrimidine
Synonyms: 2,4-dibromopyrimidine
Molecular Weight (g/mol): 237.88
Molecular Formula: C4H2Br2N2
InChi Key: AOEHEEBFRCAFGC-UHFFFAOYSA-N
SMILES: BrC1=NC(Br)=NC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results