accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BrN

CAS RN 39186-58-8

IUPAC Name: 4-bromo-2,2-diphenylbutanenitrile
Synonyms: 4-bromo-2,2-diphenylbutanenitrile
Molecular Weight (g/mol): 300.2
Molecular Formula: C16H14BrN
InChi Key: IGYSFJHVFHNOEI-UHFFFAOYSA-N
SMILES: BrCCC(C#N)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results