CAS RN 39096-01-0
CAS RN 39096-01-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N,N-Diethyl-2-hydroxyacetamide, TRC
CAS: 39096-01-0 Molecular Formula: C6H13NO2 Molecular Weight (g/mol): 131.17 Synonym: 2-Hydroxy-N,N-diethylacetamide,N,N-Diethylglycolamide,N,N-diethylglycolamide IUPAC Name: N,N-diethyl-2-hydroxyacetamide SMILES: CCN(CC)C(=O)CO