CAS RN 38930-18-6
IUPAC Name:
tripotassium hexanitrite iridiumtriuide
Synonyms:
tripotassium hexanitrite iridiumtriuide
Molecular Weight (g/mol):
585.55
Molecular Formula:
IrK3N6O12
InChi Key:
ZCAFZBZDQUBNNR-UHFFFAOYSA-H
SMILES:
[K+].[K+].[K+].[Ir-3].[O-]N=O.[O-]N=O.[O-]N=O.[O-]N=O.[O-]N=O.[O-]N=O
1
–
1
of
1
results
1
Potassium hexanitroiridate(III), Ir 32.3% min
CAS: 38930-18-6 Molecular Formula: IrK3N6O12 Molecular Weight (g/mol): 585.54 MDL Number: MFCD00049672 InChI Key: CCYKMZHCKSCCLX-UHFFFAOYSA-N Synonym: irn6o12.3k,tripotassium hexanitroiridiumtriuide,potassium hexanitroiridate iii , ir PubChem CID: 71311510 SMILES: [K+].[K+].[K+].[O-][N+](=O)[Ir+3]([N+]([O-])=O)([N+]([O-])=O)([N+]([O-])=O)([N+]([O-])=O)[N+]([O-])=O