CAS RN 389-31-1
IUPAC Name:
2-(2-fluorophenyl)-2-hydroxyacetic acid
Synonyms:
(2-fluorophenyl)(hydroxy)acetic acid
Molecular Weight (g/mol):
170.14
Molecular Formula:
C8H7FO3
InChi Key:
WWSRHIZZWWDONI-UHFFFAOYNA-N
SMILES:
OC(C(O)=O)C1=CC=CC=C1F
1
–
1
of
1
results
1
2-Fluoromandelic acid, 99%, Thermo Scientific™
CAS: 389-31-1 Molecular Formula: C8H7FO3 Molecular Weight (g/mol): 170.14 MDL Number: MFCD03095382 InChI Key: WWSRHIZZWWDONI-UHFFFAOYNA-N Synonym: 2-fluoromandelic acid,2-2-fluorophenyl-2-hydroxyacetic acid,2-fluorophenyl hydroxy acetic acid,o-fluoromandelic acid,2-fluoro-dl-mandelic acid,2-fluorophenyl hydroxyacetic acid,2-fluoro-alpha-hydroxyphenylacetic acid,2-2-fluorophenyl-2-hydroxyaceticacid,benzeneacetic acid,2-fluoro-a-hydroxy-, ar PubChem CID: 2783169 IUPAC Name: 2-(2-fluorophenyl)-2-hydroxyacetic acid SMILES: OC(C(O)=O)C1=CC=CC=C1F