accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 388582-41-0

IUPAC Name: 6-amino-2-methyl-1,4-dihydropyrimidin-4-one dihydrate
Synonyms: 6-amino-2-methyl-1H-pyrimidin-4-one dihydrate
Molecular Weight (g/mol): 161.16
Molecular Formula: C5H11N3O3
InChi Key: QJZWHXSXVBZLJI-UHFFFAOYSA-N
SMILES: O.O.CC1=NC(=O)C=C(N)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results