CAS RN 38841-98-4
IUPAC Name:
magnesium(2+) octan-1-ide chloride
Synonyms:
magnesium(2+) octan-1-ide chloride
Molecular Weight (g/mol):
172.98
Molecular Formula:
C8H17ClMg
InChi Key:
HQDAZWQQKSJCTM-UHFFFAOYSA-M
SMILES:
[Mg++].[Cl-].CCCCCCC[CH2-]
1
–
1
of
1
results
1
Octylmagnesium chloride, 1.4M solution in THF, AcroSeal™
CAS: 38841-98-4 Molecular Formula: C8H17ClMg Molecular Weight (g/mol): 172.98 MDL Number: MFCD00000476 InChI Key: DPBWWQDKVQLWFK-UHFFFAOYSA-M Synonym: octylmagnesium chloride,magnesium, chlorooctyl,n-octylmagnesiumchloride,octyl magnesium chloride,n-c8h17mgcl,n-octylmagnesium chloride,octylmagnesium chloride solution,dpbwwqdkvqlwfk-uhfffaoysa-m,1-octylmagnesium chloride, 1m in methf,octylmagnesium chloride solution, 2.0 m in thf PubChem CID: 4147387 SMILES: CCCCCCCC[Mg]Cl