accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ONNHO

CAS RN 388088-73-1

IUPAC Name: [6-(morpholin-4-yl)pyridin-3-yl]methanol
Synonyms: [6-(morpholin-4-yl)pyridin-3-yl]methanol
Molecular Weight (g/mol): 194.23
Molecular Formula: C10H14N2O2
InChi Key: NXKXCMMRCZMJRJ-UHFFFAOYSA-N
SMILES: OCC1=CN=C(C=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results