accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 38767-72-5

NHONH2

CAS RN 38767-72-5

IUPAC Name: 4-amino-1,2-dihydropyridin-2-one
Synonyms: 4-amino-1H-pyridin-2-one
Molecular Weight (g/mol): 110.12
Molecular Formula: C5H6N2O
InChi Key: SBQVQYPJWLJRQT-UHFFFAOYSA-N
SMILES: NC1=CC(=O)NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results