accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3SOOS

CAS RN 38695-60-2

IUPAC Name: 2-methanesulfonylthiophene
Synonyms: thiophene, 2-(methylsulfonyl)-
Molecular Weight (g/mol): 162.22
Molecular Formula: C5H6O2S2
InChi Key: BNYLGFINMVZZGA-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results