accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OSOOFFF

CAS RN 3857-83-8

IUPAC Name: naphthalen-2-yl trifluoromethanesulfonate
Synonyms: naphthalen-2-yl trifluoromethanesulfonate
Molecular Weight (g/mol): 276.23
Molecular Formula: C11H7F3O3S
InChi Key: MDWRQYBWVTXIIJ-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)OC1=CC2=CC=CC=C2C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results