accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NNH3CCl

CAS RN 38557-72-1

IUPAC Name: 2-chloro-3,5-dimethylpyrazine
Synonyms: 2-chloro-3,5-dimethylpyrazine
Molecular Weight (g/mol): 142.59
Molecular Formula: C6H7ClN2
InChi Key: BTGGHNHGPURMEO-UHFFFAOYSA-N
SMILES: CC1=CN=C(Cl)C(C)=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results