CAS RN 38533-61-8
IUPAC Name:
2-chloro-6-methoxy-3-nitropyridine
Synonyms:
2-chloro-6-methoxy-3-nitropyridine
Molecular Weight (g/mol):
188.57
Molecular Formula:
C6H5ClN2O3
InChi Key:
DVRGUTNVDGIKTP-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C(Cl)=N1)[N+]([O-])=O
1
–
2
of
2
results
1
2-Chloro-6-methoxy-3-nitropyridine, 98%
CAS: 38533-61-8 Molecular Formula: C6H5ClN2O3 Molecular Weight (g/mol): 188.57 MDL Number: MFCD00130268 InChI Key: DVRGUTNVDGIKTP-UHFFFAOYSA-N Synonym: 2-chloro-3-nitro-6-methoxypyridine,pyridine, 2-chloro-6-methoxy-3-nitro,2-chloro-6-methoxy-3-nitro-pyridine,2-chloro-6-methoxy-3 or 5-nitropyridine,pubchem4044,acmc-1agmz,ksc226c7t,2-chloro-3nitro-6-methoxy-pyridine,6-methoxy-2-chloro-3-nitropyridine,2-chloro-6-methoxy-3-nitropyridine PubChem CID: 2795029 IUPAC Name: 2-chloro-6-methoxy-3-nitropyridine SMILES: COC1=CC=C(C(Cl)=N1)[N+]([O-])=O