accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 384346-91-2

IUPAC Name: tert-butyl({[2-(4-methoxyphenyl)-2H,4H,4aH,8H,8aH-pyrano[3,2-d][1,3]dioxin-8-yl]oxy})dimethylsilane
Synonyms: tert-butyl({[2-(4-methoxyphenyl)-2H,4H,4aH,8H,8aH-pyrano[3,2-d][1,3]dioxin-8-yl]oxy})dimethylsilane
Molecular Weight (g/mol): 378.54
Molecular Formula: C20H30O5Si
InChi Key: ZTRSSQRWGPRALW-UHFFFAOYNA-N
SMILES: COC1=CC=C(C=C1)C1OCC2OC=CC(O[Si](C)(C)C(C)(C)C)C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results