accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NHOCl

CAS RN 384346-27-4

IUPAC Name: 3-(4-chlorophenoxy)piperidine
Synonyms: 3-(4-chlorophenoxy)piperidine
Molecular Weight (g/mol): 211.69
Molecular Formula: C11H14ClNO
InChi Key: OXELOOCWZRBKSV-UHFFFAOYNA-N
SMILES: ClC1=CC=C(OC2CCCNC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results