accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 38401-67-1

H3COSHO

CAS RN 38401-67-1

IUPAC Name: 1-[5-(4-methoxyphenyl)thiophen-2-yl]ethan-1-one
Synonyms: 1-[5-(4-methoxyphenyl)thiophen-2-yl]ethanone
Molecular Weight (g/mol): 232.3
Molecular Formula: C13H12O2S
InChi Key: LRUBIGCXVSIWPI-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=CC=C(S1)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results