accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CONHNH2

CAS RN 38291-82-6

IUPAC Name: pentanehydrazide
Synonyms: valeric acid hydrazide
Molecular Weight (g/mol): 116.16
Molecular Formula: C5H12N2O
InChi Key: PJBQYCIDGYKEMN-UHFFFAOYSA-N
SMILES: CCCCC(=O)NN
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results