accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 38289-29-1

H3COHO

CAS RN 38289-29-1

IUPAC Name: 4-pentylcyclohexane-1-carboxylic acid
Synonyms: 4-pentylcyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 198.31
Molecular Formula: C12H22O2
InChi Key: RVLAXPQGTRTHEV-UHFFFAOYSA-N
SMILES: CCCCCC1CCC(CC1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results