accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OONNSCH3

CAS RN 38275-41-1

IUPAC Name: methyl 2-(methylsulfanyl)pyrimidine-5-carboxylate
Synonyms: methyl 2-(methylsulfanyl)pyrimidine-5-carboxylate
Molecular Weight (g/mol): 184.21
Molecular Formula: C7H8N2O2S
InChi Key: PSYRMEZGAWNWHV-UHFFFAOYSA-N
SMILES: COC(=O)C1=CN=C(SC)N=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)