accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 38245-36-2

OOOOOOOOOGdGd

CAS RN 38245-36-2

IUPAC Name: digadolinium(3+) hydrate tricarbonate
Synonyms: digadolinium(3+) hydrate tricarbonate
Molecular Weight (g/mol): 512.54
Molecular Formula: C3H2Gd2O10
InChi Key: MXYFVHNBWNHETM-UHFFFAOYSA-H
SMILES: O.[Gd+3].[Gd+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)