accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 38222-83-2

CH3NH3CCH3H3CCH3H3CCH3

CAS RN 38222-83-2

IUPAC Name: 2,6-di-tert-butyl-4-methylpyridine
Synonyms: 2,6-di-tert-butyl-4-methylpyridine
Molecular Weight (g/mol): 205.35
Molecular Formula: C14H23N
InChi Key: HVHZEKKZMFRULH-UHFFFAOYSA-N
SMILES: CC1=CC(=NC(=C1)C(C)(C)C)C(C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (3)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)