accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 38215-36-0

H3CNCH3OOSN

CAS RN 38215-36-0

IUPAC Name: 3-(1,3-benzothiazol-2-yl)-7-(diethylamino)-2H-chromen-2-one
Synonyms: coumarin 6
Molecular Weight (g/mol): 350.44
Molecular Formula: C20H18N2O2S
InChi Key: VBVAVBCYMYWNOU-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C2C=C(C3=NC4=CC=CC=C4S3)C(=O)OC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)