accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 38076-80-1

OOHNOCl

CAS RN 38076-80-1

IUPAC Name: 5-chloro-2-oxo-1,2-dihydropyridine-3-carboxylate
Synonyms: 5-chloro-2-oxo-1H-pyridine-3-carboxylate
Molecular Weight (g/mol): 172.54
Molecular Formula: C6H3ClNO3
InChi Key: XBHXNMLFJZTSAS-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(Cl)=CNC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results