accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BrSOBr

CAS RN 38071-22-6

IUPAC Name: 4,5-dibromothiophene-2-carbaldehyde
Synonyms: 4,5-dibromothiophene-2-carbaldehyde
Molecular Weight (g/mol): 269.94
Molecular Formula: C5H2Br2OS
InChi Key: CHRAVDVWAPGZOY-UHFFFAOYSA-N
SMILES: BrC1=C(Br)C=C(S1)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results