accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OPOSOCH3

CAS RN 38066-16-9

IUPAC Name: diethyl [(phenylsulfanyl)methyl]phosphonate
Synonyms: diethyl (phenylsulfanyl)methylphosphonate
Molecular Weight (g/mol): 260.29
Molecular Formula: C11H17O3PS
InChi Key: FBUXEPJJFVDUFE-UHFFFAOYSA-N
SMILES: CCOP(=O)(CSC1=CC=CC=C1)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results