accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 380594-11-6

NOOClOOCl

CAS RN 380594-11-6

IUPAC Name: 5-(2-chloro-4-nitrophenyl)furan-2-carbonyl chloride
Synonyms: 5-(2-chloro-4-nitrophenyl)furan-2-carbonyl chloride
Molecular Weight (g/mol): 286.06
Molecular Formula: C11H5Cl2NO4
InChi Key: BQTYBZIOLAXUSL-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=C(C=C1)C1=CC=C(O1)C(Cl)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results