accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H2NO(S)OH(S)OH(S)OH(R)OHO

CAS RN 3789-97-7

IUPAC Name: (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanamide
Synonyms: glucuronamide
Molecular Weight (g/mol): 193.16
Molecular Formula: C6H11NO6
InChi Key: JRIOKBXQMHEJOZ-QTBDOELSSA-N
SMILES: NC(=O)[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results