accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CH3CCH3OONHOHO

CAS RN 37736-82-6

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-cyclohexylpropanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-cyclohexylpropanoic acid
Molecular Weight (g/mol): 271.36
Molecular Formula: C14H25NO4
InChi Key: MSZQAQJBXGTSHP-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC1CCCCC1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results